Victoria blue R, Sigma-Aldrich®
Proveedor: SIGMA-ALDRICH MICROSCOPY
M-Clarity™ quality level = MQ100
234176-25GEA
111
EUR
234176-25G
Victoria blue R, Sigma-Aldrich®
Victoria blue R
Formula:
C₂₉H₃₂ClN₃ |
Número MDL:
MFCD00011876 Núm. CAS: 2185-86-6 EINECS: 218-572-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...