Fraction collection tubes, Chromacol™
Proveedor: Thermo Scientific
CRMAFRACG-13100EA 56.7 EUR
CRMAFRACG-13100 CRMAFRAC-16100
Fraction collection tubes, Chromacol™
Borosilicate glass. Capable of dealing with the demands of preparative liquid fraction collection equipment and liquid handling systems.
  • Base is precision formed to give a stable and secure position in the chosen racks
  • Wall thickness to give the robustness for routine use and collection
  • Reproducible recovery of compounds, whether acidic or basic
Expansion versions are re-usable and may be used in a range of drying, heating and evaporation instruments.