Picolinamide ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAP0414-25GEA
92
EUR
TCIAP0414-25G
Picolinamide ≥98.0% (by HPLC, titration analysis)
Picolinamide
Formula:
C₆H₆N₂O MW: 122,13 g/mol Pto. Ebullición: 143°C |
Número MDL:
MFCD00023483 Núm. CAS: 1452-77-3 EINECS: 215-921-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...