4-Bromo-4'-heptylbiphenyl ≥97.0% (por GC)
Proveedor: TCI EUROPE
TCIAB1304-5GEA
137
EUR
TCIAB1304-5G
4-Bromo-4'-heptylbiphenyl ≥97.0% (por GC)
4-Bromo-4'-heptylbiphenyl
Formula:
C₁₉H₂₃Br MW: 331,2899 g/mol |
Número MDL:
MFCD00060107 Núm. CAS: 58573-93-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 92.0 to 95.0 deg-C |
Solubility in Toluene | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...