Magnesium tantalate ≥99.9% (metals basis)

39366.14EA 153 EUR
39366.14
Magnesium tantalate ≥99.9% (metals basis)
Magnesium tantalate
Formula: MgO₆Ta₂
Número MDL: MFCD00210633
Núm. CAS: 12293-61-7
EINECS: 235-562-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR