(+)-DIOP ≥95.0% (by GC, titration analysis)
|
Atención
|
TCIAB1112-1GEA
122
EUR
TCIAB1112-1G
(+)-DIOP ≥95.0% (by GC, titration analysis)
(+)-DIOP
Formula:
C₃₁H₃₂O₂P₂ MW: 498,532 g/mol |
Número MDL:
MFCD00009760 Núm. CAS: 37002-48-5 EINECS: 253-307-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Iodometric Titration) | min. 95.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...