3,5-Dichloro-4-hydroxybenzonitrile, Supelco®

Proveedor: Merck


M-Clarity™ quality level = MQ100

33363-25MGEA 82.6 EUR
33363-25MG
3,5-Dichloro-4-hydroxybenzonitrile, Supelco®
3,5-Dichloro-4-hydroxybenzonitrile
Formula: C₇H₃Cl₂NO
Número MDL: MFCD00002177
Núm. CAS: 1891-95-8
EINECS: 217-572-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR