3,5-Dichloro-4-hydroxybenzonitrile
Proveedor: EHRENSTORFER GMBH
EHERC11550000EA
159
EUR
EHERC11550000
3,5-Dichloro-4-hydroxybenzonitrile
3,5-Dichloro-4-hydroxybenzonitrile
Formula:
C₇H₃Cl₂NO |
Número MDL:
MFCD00002177 Núm. CAS: 1891-95-8 EINECS: 217-572-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...