Probenecid
Proveedor: LGC Standards PROMOCHEM
PROOMM0657.00EA
1106
EUR
PROOMM0657.00
Probenecid
Probenecid
Formula:
C₁₃H₁₉NO₄S MW: 285,4 g/mol |
Número MDL:
MFCD00038402 Núm. CAS: 57-66-9 EINECS: 200-344-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...