Quinoxaline-2,3-dione ≥98.0% (by titrimetric analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAD1010-25GEA
43
EUR
TCIAD1010-25G
Quinoxaline-2,3-dione ≥98.0% (by titrimetric analysis)
Quinoxaline-2,3-dione
Formula:
C₈H₆N₂O₂ MW: 162,15 g/mol |
Número MDL:
MFCD00006723 Núm. CAS: 15804-19-0 EINECS: 239-901-0 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystaline |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...