Okadaic acid

Proveedor: Enzo Life Sciences

ALEXALX350003C025EA 123 EUR
ALEXALX350003C025 ALEXALX350003-M001 ALEXALX350003C050 ALEXALX350003-C100
Okadaic acid
Okadaic acid
Formula: C₄₄H₆₈O₁₃
MW: 805,02 g/mol
Número MDL: MFCD00083455
Núm. CAS: 78111-17-8
https://es.vwr.com/stibo/web/std.lang.all/76/41/45347641.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR