Diethylzinc ∼15% ∼1 mol/L in toluene
Proveedor: TCI EUROPE
|
Peligro
|
TCIAD3902-100MLEA
62
EUR
TCIAD3902-100ML
Diethylzinc ∼15% ∼1 mol/L in toluene
Diethylzinc
Formula:
C₄H₁₀Zn MW: 123,51 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00009021 Núm. CAS: 557-20-0 EINECS: 209-161-3 UN: 3399 ADR: 4.3,I |
Resultados test especificaciones
Appearance | Colorless to Light yellow clear liquid to cloudy liquid |
Concentration (Chelometric Titration) | 14.5 to 16.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...