Crafts jacket, FAS-459
Proveedor: Fristads Kansas Group
KWIN100305940LEA 68.9 EUR
KWIN100305940L KWIN100305940XS KWIN1003059403XL KWIN1003059402XL KWIN100305940XL KWIN100305940S KWIN100305940M
Crafts jacket, FAS-459
Ropa de trabajo
Versatile jacket made of FAS® twill (100% cotton).
  • Zip to top of collar, ventilated back
  • Adjustable cuffs and hem
  • Security pocket, two front pockets
  • Two chest pockets with flap and button, one with button and loop for sheath knife, one with tool loops
  • Concealed ID-card pocket, sleeve pocket, inside pocket with button
  • Inside mobile phone pocket, earpiece loop
  • ÖKO-TEX® certified
Weight: 375 g/m²