Flasks, pear shaped, Claisen-Vigreux
Proveedor: SciLabware
BIBBMD4/50EA 121 EUR
BIBBMD4/50 BIBBMD4/100
Flasks, pear shaped, Claisen-Vigreux
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Incorporates a short, Vigreux type fractionating column. Column indents aid intimate contact between vapour and liquid.