(R)-(-)-2-Heptyl isothiocyanate ≥98%
Proveedor: Thermo Scientific
|
Peligro
|
L20064.03EA
94.3
EUR
L20064.03
(R)-(-)-2-Heptyl isothiocyanate ≥98%
(R)-(-)-2-Heptyl isothiocyanate
Formula:
C₈H₁₅NS MW: 157,28 g/mol Densidad: 0,910 |
Número MDL:
MFCD05664051 Núm. CAS: 737000-93-0 EINECS: 000-000-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...