tert-Butylhydrazine hydrochloride 98%
Proveedor: Thermo Scientific
|
Atención
|
ACRO187081000EA
127
EUR
ACRO187081000
tert-Butylhydrazine hydrochloride 98%
tert-Butylhydrazine hydrochloride
Formula:
C₄H₁₃ClN₂ MW: 124,61 g/mol Pto. de fusión: 190…194 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00012947 Núm. CAS: 7400-27-3 EINECS: 231-001-6 |
Resultados test especificaciones
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 190 to 194 °C |
Titration with HClO4 | ≥97.5 % |
Additional info | Denatured with 1% iPr-OH, 1% MEK and 0.001% Bitrex |
Ethanol | 99.8 to 100.0 % v/v (calculated) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...