4-Pyrid-3-ylbenzoic acid

Proveedor: Thermo Scientific

MAYBCC40001DAEA 0 EUR
MAYBCC40001DA
4-Pyrid-3-ylbenzoic acid
4-Pyrid-3-ylbenzoic acid
Formula: C₁₂H₉NO₂
MW: 199,2054 g/mol
Número MDL: MFCD03426513
Núm. CAS: 4385-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR