2-Propylimidazole ≥95.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAP0947-5GEA
42
EUR
TCIAP0947-5G
TCIAP0947-25G
2-Propylimidazole ≥95.0% (by GC, titration analysis)
2-Propylimidazole
Formula:
C₆H₁₀N₂ MW: 110,1570 g/mol Pto. Ebullición: 152°C |
Número MDL:
MFCD00059158 Núm. CAS: 50995-95-4 EINECS: 256-902-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 56.0 to 61.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...