(S)-(+)-Glycidyl butyrate ≥98%, ee 98%
Proveedor: Thermo Scientific
L18558.03EA
37.4
EUR
L18558.03
(S)-(+)-Glycidyl butyrate ≥98%, ee 98%
(S)-(+)-Glycidyl butyrate
Formula:
C₇H₁₂O₃ MW: 144,1684 g/mol Pto. de inflamación: 85 °C (185 °F) |
Número MDL:
MFCD00273369 Núm. CAS: 65031-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...