Tetrakis(dimethylamino)zirconium(IV)
|
Peligro
|
ACRO380410050EA
195
EUR
ACRO380410050
ACRO380410010
Tetrakis(dimethylamino)zirconium(IV)
Tetrakis(dimethylamino)zirconium(IV)
Formula:
C₈H₂₄N₄Zr MW: 267,527 g/mol Pto. de fusión: 57…60 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00239502 Núm. CAS: 19756-04-8 UN: 3396 ADR: 4.3,II |
Resultados test especificaciones
Appearance | Yellow-green Crystalline solid |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...