3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt ≥99.0% (by titrimetric analysis) for biochemical research
Proveedor: TCI EUROPE
|
Atención
|
TCIAD1928-25GEA
96
EUR
TCIAD1928-25G
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt ≥99.0% (by titrimetric analysis) for biochemical research
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt
Formula:
C₆H₃Cl₂NaO₄S MW: 265,05 g/mol |
Número MDL:
MFCD00009798 Núm. CAS: 54970-72-8 EINECS: 259-416-8 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Ion exchange titration) | min. 99.0 % |
Hydrogendioxide detection test (Peroxidase method) | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...