Di-tert-butylphenylphosphonium tetrafluoroborate ≥99%
Proveedor: Thermo Scientific
|
Atención
|
H26904.03EA
380
EUR
H26904.03
Di-tert-butylphenylphosphonium tetrafluoroborate ≥99%
Di-tert-butylphenylphosphonium tetrafluoroborate
Formula:
C₁₄H₂₄BF₄P MW: 310,12 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08704553 Núm. CAS: 612088-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...