Tetraphenylphosphonium tetraphenylborate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI EUROPE

TCIAT1329-25GEA 70 EUR
TCIAT1329-25G
Tetraphenylphosphonium tetraphenylborate ≥98.0% (by HPLC, titration analysis)
Tetraphenylphosphonium tetraphenylborate
Formula: C₄₈H₄₀BP
MW: 658,6160 g/mol
Número MDL: MFCD00145389
Núm. CAS: 15525-15-2
https://es.vwr.com/stibo/web/std.lang.all/53/60/28995360.jpg

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of Ion-Pair)
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR