Storage cooled incubators, IPS series
Proveedor: Memmert
MEMMIPS750EA 0 EUR
MEMMIPS750
Storage cooled incubators, IPS series
Incubadoras Incubadoras refrigeradas
The IPS cooled storage incubators are used for storing samples at constant temperatures which meet the requirements of the food, cosmetics, medicine and pharmaceutical industries.
  • Temperature range 14 to 45 °C
  • Double doors prevent contamination and temperature drops
  • Integrated fans ensure rapid energy transfer and optimal temperature distribution
  • Intuitive and easy to use operating menu
  • Wide range of options for programming and documentation using interfaces, integrated data logger and AtmoCONTROL software
Both the chamber and housing of the units are made from corrosion-resistant stainless steel which is easily cleaned. The heating concept is specifically adapted for precise and homogenous temperature control.