1-Cloro-4-etoxibenceno ≥99.0% (por GC)
Proveedor: TCI EUROPE
TCIAC0239-25GEA
112
EUR
TCIAC0239-25G
1-Cloro-4-etoxibenceno ≥99.0% (por GC)
1-Cloro-4-etoxibenceno
Formula:
C₈H₉ClO MW: 156,61 g/mol Pto. Ebullición: 214°C |
Número MDL:
MFCD00058931 Núm. CAS: 622-61-7 EINECS: 210-747-6 |
![]() |
Resultados test especificaciones
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Freezing point | 16.0 to 19.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...