Kaempferol

Proveedor: Enzo Life Sciences

ALEXALX385005M050EA 189 EUR
ALEXALX385005M050 ALEXALX385005M010
Kaempferol
Kaempferol
Formula: C₁₅H₁₀O₆
MW: 286,24 g/mol
Número MDL: MFCD00016938
Núm. CAS: 520-18-3
EINECS: 208-287-6
https://es.vwr.com/stibo/web/std.lang.all/05/74/45390574.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR