4-Amino-3,5-xylenol ≥98.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAA1860-25GEA
93
EUR
TCIAA1860-25G
4-Amino-3,5-xylenol ≥98.0% (by GC, titration analysis)
4-Amino-3,5-xylenol
Formula:
C₈H₁₁NO MW: 137,1800 g/mol |
Número MDL:
MFCD01741459 Núm. CAS: 3096-70-6 EINECS: 221-448-5 |
![]() |
Resultados test especificaciones
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 180.0 to 186.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...