Hexadecyl chloroformate ≥98.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Peligro
|
TCIAC1053-25MLEA
51
EUR
TCIAC1053-25ML
Hexadecyl chloroformate ≥98.0% (by GC, titration analysis)
Hexadecyl chloroformate
Formula:
C₁₇H₃₃ClO₂ MW: 304,8957 g/mol |
Número MDL:
MFCD00035954 Núm. CAS: 26272-90-2 EINECS: 247-578-2 UN: 3277 ADR: 6.1,II |
![]() |
Resultados test especificaciones
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...