Mesitylacetic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI EUROPE
Atención

TCIAM1218-5GEA 121 EUR
TCIAM1218-5G
Mesitylacetic acid ≥98.0% (by titrimetric analysis)
Mesitylacetic acid
Formula: C₁₁H₁₄O₂
MW: 178,23 g/mol
Número MDL: MFCD00014368
Núm. CAS: 4408-60-0
EINECS: 224-556-0
https://es.vwr.com/stibo/web/std.lang.all/49/98/28974998.jpg

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR