4-(Trifluoromethyl)benzoic anhydride ≥97.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAT1593-10GEA
134
EUR
TCIAT1593-10G
4-(Trifluoromethyl)benzoic anhydride ≥97.0% (by GC, titration analysis)
4-(Trifluoromethyl)benzoic anhydride
Formula:
C₁₆H₈F₆O₃ MW: 362,23 g/mol |
Número MDL:
MFCD00671577 Núm. CAS: 25753-16-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Morpholine Method) | min. 97.0 % |
Melting point | 127.0 to 133.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...