Benzophenone-2,4'-dicarboxylic acid monohydrate ≥98.0% (by titrimetric analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAB0084-25GEA
110
EUR
TCIAB0084-25G
Benzophenone-2,4'-dicarboxylic acid monohydrate ≥98.0% (by titrimetric analysis)
Benzophenone-2,4'-dicarboxylic acid monohydrate
Formula:
C₁₅H₁₀O₅·1H₂O |
Número MDL:
MFCD00016508 Núm. CAS: 85-58-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0% (calcd.on anh.substance) |
Solubility in Methanol | almost transparency |
Water | 5.6 to 6.6 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...