5-Bromo-2-hydroxybenzophenone ≥98.0% (by titrimetric analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAB2341-5GEA
130
EUR
TCIAB2341-5G
5-Bromo-2-hydroxybenzophenone ≥98.0% (by titrimetric analysis)
5-Bromo-2-hydroxybenzophenone
Formula:
C₁₃H₉BrO₂ MW: 277,11 |
Número MDL:
MFCD00525062 Núm. CAS: 55082-33-2 |
![]() |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 109.0 to 114.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...