2-(Methylsulphonyl)acetophenone ≥98.0% (por GC)
Proveedor: TCI EUROPE
TCIAM0712-5GEA
144
EUR
TCIAM0712-5G
2-(Methylsulphonyl)acetophenone ≥98.0% (por GC)
2-(Methylsulphonyl)acetophenone
Formula:
C₉H₁₀O₃S |
Número MDL:
MFCD00025078 Núm. CAS: 3708-04-1 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 108.0 to 112.0 deg-C |
Solubility in hot EtOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...