2-(Isopropylamino)acetophenone hydrochloride 98%
Proveedor: Thermo Scientific
|
Atención
|
H50799.MDEA
76.1
EUR
H50799.MD
2-(Isopropylamino)acetophenone hydrochloride 98%
2-(Isopropylamino)acetophenone hydrochloride
Formula:
C₁₁H₁₆ClNO MW: 213,71 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD18785640 Núm. CAS: 38506-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...