Ergocalciferol (vitamin D2, calciferol) ≥98.0% (by HPLC)
Proveedor: TCI EUROPE
|
Peligro
|
TCIAC0005-1GEA
53
EUR
TCIAC0005-1G
Ergocalciferol (vitamin D2, calciferol) ≥98.0% (by HPLC)
Ergocalciferol (vitamin D2, calciferol)
Formula:
C₂₈H₄₄O MW: 396,66 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00166988 Núm. CAS: 50-14-6 EINECS: 200-014-9 UN: 2811 ADR: 6.1,II |
Resultados test especificaciones
Appearance | White powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 111.0 to 116.0 deg-C |
Specific rotation [a]20/D | +100 to +109 deg (C=1.5, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...