Hydrochlorothiazide

Proveedor: EHRENSTORFER GMBH

C14223500
EHERC14223500EA 189 EUR
EHERC14223500
Hydrochlorothiazide
Hydrochlorothiazide
Formula: C₇H₈ClN₃O₄S₂
Número MDL: MFCD00051765
Núm. CAS: 58-93-5
EINECS: 200-403-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR