Folic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI EUROPE
TCIAF0043-25GEA
58
EUR
TCIAF0043-25G
Folic acid ≥98.0% (by HPLC, titration analysis)
Folic acid
Formula:
C₁₉H₁₉N₇O₆ MW: 441,41 g/mol |
Número MDL:
MFCD00079305 Núm. CAS: 59-30-3 EINECS: 200-419-0 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in NaOH (100g/L) | almost transparency |
Water | max. 8.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...