Folic acid

Proveedor: Enzo Life Sciences

ALEXALX460006G010EA 47 EUR
ALEXALX460006G010
Folic acid
Folic acid
Formula: C₁₉H₁₉N₇O₆
MW: 441,41 g/mol
Número MDL: MFCD00079305
Núm. CAS: 59-30-3
EINECS: 200-419-0
https://es.vwr.com/stibo/web/std.lang.all/07/48/45390748.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR