Diclofenac sodium salt
Proveedor: LGC Standards PROMOCHEM
PROOBP619EA
288
EUR
PROOBP619
Diclofenac sodium salt
Diclofenac sodium salt
Formula:
C₁₄H₁₀Cl₂NNaO₂ MW: 318,1 g/mol |
Número MDL:
MFCD00082251 Núm. CAS: 15307-79-6 EINECS: 239-346-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...