Thiodicarb, Supelco®

Proveedor: Merck


M-Clarity™ quality level = MQ100

34375-250MGEA 114 EUR
34375-250MG
Thiodicarb, Supelco®
Thiodicarb
Formula: C₁₀H₁₈N₄O₄S₃
Número MDL: MFCD00145401
Núm. CAS: 59669-26-0
EINECS: 261-848-7
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR