Portable sampler, PB 13
Proveedor: WTW
WTWO000184EA 5140 EUR
WTWO000184
Portable sampler, PB 13
Portable sampler PB 13 with vacuum technology for universal applications. PB 13 can be mounted directly on all common storage containers or onto the IsoBox, which is available in different versions.
  • Low maintenance vacuum system
  • Large number of sample cycles with just one battery charge
  • Sampling is traceable on display
When combined with IsoBox, it provides the user with a practical and modularly extendable system with passive cooling. IsoBox is not only suitable for temperature control of samples but also for safe protected sample transport.
Portable microprocessor-controlled liquid sampler with vacuum dosing system and built-in rechargeable battery.
Información de suministro: Delivery includes 5 m suction tubing, screw fitting and tubing end piece. battery charger, sample containers or IsoBox are not included.