Staining jars acc. to Coplin
Proveedor: Marienfeld
MARI5600003EA 77.9 EUR
MARI5600003
Staining jars acc. to Coplin
Made of PP, with screw-cap.
  • Break-resistant
  • Good chemical resistance
  • Good temperature resistance
These staining jar hold up to 10 slides (76×26 mm).