DNA labelling, Amersham Rediprime II™ DNA Labelling System
Proveedor: Cytiva
Rediprime™
RPN1633EA 561 EUR
RPN1633
DNA labelling, Amersham Rediprime II™ DNA Labelling System
Reactivos para electroforesis Reactivos para Blotting
Rediprime II™ DNA Labelling System consists of premixed, individually dispensed, random-prime reactions, freeze-dried for ambient stability.
Simply add substrate and [α-³²P]dCTP. As little as 25 ng substrate DNA can be used. Suitable for use with dCTP only.
Información de suministro: Each system includes 30X labelling mix (buffered solution of dATP, dGTP, dTTP, exonuclease free Klenow enzyme and random primers in a dried, stabilised form) and Control DNA (lambda Hind III DNA in a dried, stabilised form).