Water Jet Pumps, MBL®, SciLabware
Proveedor: SciLabware
BIBBFRF442EA 297 EUR
BIBBFRF442
Water Jet Pumps, MBL®, SciLabware
Water jet pumps made of chemically resistant borosilicate glass.
  • Generates a moderate vacuum (up to 65 mbar) when connected to a water supply
  • With a check valve to prevent back-suction and waterlogging