Safety waste disposal system
Proveedor: Vitlab
VITL151594EA 260 EUR
VITL151594
Safety waste disposal system
Frascos Botellas de plástico
PE/PP.
  • Good chemical resistance
  • Good temperature resistance
For collection of liquid chemicals in the laboratory. The inlet hopper made from HDPE contains a self-closing float, overfill protection, and a splash guard.
Información de suministro: Screw cap (GL 63) with sealing ring is included.