ALFAJ62399

J62399.APEA 129 EUR
ALFAJ62399.AP
ALFAJ62399
CAS No.: 56-81-5
Molecular Formula: C3H8O3
Formula Weight: 92.09
Flash Point: 160°(320°F)
Physical Form: Viscous liquid
Density: 1.26
Refractive Index: 1.4740
Appearance: Colourless
Odour: Odourless
Melting Point: 18°
Boiling Point: 182°/20mm
Solubility: Miscible with water
Beilstein Reference: 635685
Merck Reference: 14,4484
MDL No.: MFCD00004722

Useful as a cryoprotectant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR