1,4-Bis(5-fenil-4-metiloxazol-2-il)benceno ≥99.0% (by HPLC, total nitrogen) for scintillation spectrometry
Proveedor: TCI EUROPE
Sinónimos:
Dimetil-POPOP
TCIAB0499-25GEA
330
EUR
TCIAB0499-25G
1,4-Bis(5-fenil-4-metiloxazol-2-il)benceno ≥99.0% (by HPLC, total nitrogen) for scintillation spectrometry
1,4-Bis(5-fenil-4-metiloxazol-2-il)benceno
Formula:
C₂₆H₂₀N₂O₂ MW: 392,46 g/mol Pto. de fusión: 234…235 °C |
Número MDL:
MFCD00005310 Núm. CAS: 3073-87-8 EINECS: 221-349-7 |
Resultados test especificaciones
Appearance | White to Light yellow to Green powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (with Total Nitrogen) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...