2-Mercapto-5-methoxybenzothiazole ≥98.0% (by titrimetric analysis)

Proveedor: TCI EUROPE
Atención

TCIAM1277-5GEA 195 EUR
TCIAM1277-5G
2-Mercapto-5-methoxybenzothiazole ≥98.0% (by titrimetric analysis)
2-Mercapto-5-methoxybenzothiazole
Formula: C₈H₇NOS₂
MW: 197,28 g/mol
Número MDL: MFCD00185941
Núm. CAS: 55690-60-3
EINECS: 259-755-1
https://es.vwr.com/stibo/web/std.lang.all/51/21/28975121.jpg

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 196.0 to 199.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR