2,4-Xylidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAX0028-25GEA
295
EUR
TCIAX0028-25G
2,4-Xylidine hydrochloride ≥98.0% (by HPLC, titration analysis)
2,4-Xylidine hydrochloride
Formula:
C₈H₁₂ClN MW: 157,6406 g/mol |
Número MDL:
MFCD00060209 Núm. CAS: 21436-96-4 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...