Dibenzenesulphonimide ≥98.0% (by HPLC)
Proveedor: TCI EUROPE
TCIAD2540-5GEA
54
EUR
TCIAD2540-5G
TCIAD2540-25G
Dibenzenesulphonimide ≥98.0% (by HPLC)
Dibenzenesulphonimide
Formula:
C₁₂H₁₁NO₄S₂ |
Número MDL:
MFCD00025019 Núm. CAS: 2618-96-4 EINECS: 220-051-4 |
![]() |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 156.0 to 160.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...